Assay Details
Target Gene Details
Entrez Gene ID: | 3892 |
Gene Name: | keratin 86 |
Gene Aliases: |
HB6, Hb1, K86, KRTHB1, KRTHB6, MNX |
Location: |
Chr.12:52270507-52309163 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT86 | NM_001320198.1 | NP_001307127.1 | ||
| XM_005268866.4 | XP_005268923.1 | |||
| XM_017019296.1 | XP_016874785.1 | |||
| AK292237.1 | ||||
| AK297794.1 | ||||
| BC069585.1 | AAH69585.1 | |||
| X99142.1 | CAA67579.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1503n100 | Chr.12:52285116 - 52394255 on Build GRCh38 | Gain |
|
| esv2759899 | Chr.12:52169480 - 52391735 on Build GRCh38 | Gain |
|
| dgv29e180 | Chr.12:52291976 - 52389509 on Build GRCh38 | Gain |
|
| nsv516938 | Chr.12:52283925 - 52394228 on Build GRCh38 | Gain+Loss |
|
| dgv487e212 | Chr.12:52294699 - 52388067 on Build GRCh38 | Gain |
|
| dgv200n27 | Chr.12:52297319 - 52395144 on Build GRCh38 | Gain |
|
| dgv9e209 | Chr.12:52294216 - 52390216 on Build GRCh38 | Duplication |
|
| dgv2630n54 | Chr.12:52287930 - 52395144 on Build GRCh38 | Gain |
|
| esv27319 | Chr.12:52287366 - 52389709 on Build GRCh38 | Gain+Loss |
|
| dgv70e55 | Chr.12:52282619 - 52398149 on Build GRCh38 | Gain |
|
| esv2760832 | Chr.12:52294900 - 52389052 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73111108] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map