Assay Details
Target Gene Details
Entrez Gene ID: | 196528 |
Gene Name: | AT-rich interaction domain 2 |
Gene Aliases: |
BAF200, p200 |
Location: |
Chr.12:45729683-45908037 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARID2 | NM_152641.2 | NP_689854.2 | ||
| XM_006719272.3 | XP_006719335.1 | |||
| AK127872.1 | BAC87171.1 | |||
| AY727870.2 | AAU20329.2 | |||
| CR749833.1 | CAH18689.1 | |||
| DQ096628.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759897 | Chr.12:45755555 - 45956108 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs117358133] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map