Assay Details
Target Gene Details
Entrez Gene ID: | 144404 |
Gene Name: | transmembrane protein 120B |
Gene Aliases: |
- |
Location: |
Chr.12:121712752-121782070 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM120B | NM_001080825.2 | NP_001074294.2 | ||
| XM_017018853.1 | XP_016874342.1 | |||
| XM_017018854.1 | XP_016874343.1 | |||
| AK126568.1 | ||||
| BC127768.1 | ||||
| BC127769.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv2919n54 | Chr.12:121739416 - 121747940 on Build GRCh38 | Loss |
|
| nsv560459 | Chr.12:121729189 - 121747874 on Build GRCh38 | Loss |
|
| esv3630957 | Chr.12:121739879 - 121746290 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79213708] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map