Assay Details
Target Gene Details
Entrez Gene ID: | 8638 |
Gene Name: | 2'-5'-oligoadenylate synthetase like |
Gene Aliases: |
OASLd, TRIP-14, TRIP14, p59 OASL, p59-OASL, p59OASL |
Location: |
Chr.12:121018882-121039242 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OASL | NM_001261825.1 | NP_001248754.1 | ||
| NM_003733.3 | NP_003724.1 | |||
| NM_198213.2 | NP_937856.1 | |||
| XM_017020140.1 | XP_016875629.1 | |||
| XM_017020141.1 | XP_016875630.1 | |||
| AJ225089.1 | CAA12396.1 | |||
| AK023956.1 | ||||
| AK314419.1 | ||||
| BC117408.1 | ||||
| BC117410.1 | ||||
| BM909951.1 | ||||
| BM918192.1 | ||||
| JQ792168.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1039216 | Chr.12:120983898 - 121100688 on Build GRCh38 | Gain |
|
| nsv560454 | Chr.12:121019270 - 121307939 on Build GRCh38 | Loss |
|
| nsv521928 | Chr.12:120603925 - 121356945 on Build GRCh38 | Gain |
|
| esv2759917 | Chr.12:120978042 - 121147010 on Build GRCh38 | Loss |
|
| nsv832531 | Chr.12:121002692 - 121166429 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map