Assay Details
Target Gene Details
Entrez Gene ID: | 11113 |
Gene Name: | citron rho-interacting serine/threonine kinase |
Gene Aliases: |
CRIK, STK21 |
Location: |
Chr.12:119685790-119877318 on Build GRCh38 |
Assay Gene Location: | Within Intron 48 |
More Information
Set Membership: |
|

Genomic Map