Assay Details
Target Gene Details
Entrez Gene ID: | 3855 |
Gene Name: | keratin 7 |
Gene Aliases: |
CK7, K2C7, K7, SCL |
Location: |
Chr.12:52233170-52252667 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT7 | NM_005556.3 | NP_005547.3 | ||
| XM_011538325.1 | XP_011536627.1 | |||
| XM_017019294.1 | XP_016874783.1 | |||
| AF509887.1 | AAN64031.1 | |||
| AJ238246.1 | CAB41416.1 | |||
| AK025881.1 | ||||
| AK128505.1 | ||||
| AK128848.1 | ||||
| AK129924.1 | ||||
| AK225498.1 | ||||
| BC002700.2 | AAH02700.1 | |||
| BC009521.1 | AAH09521.1 | |||
| BE740515.1 | ||||
| X03212.1 | CAA26956.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759899 | Chr.12:52169480 - 52391735 on Build GRCh38 | Gain |
|
| dgv1501n100 | Chr.12:52237529 - 52281537 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map