Assay Details
Target Gene Details
Entrez Gene ID: | 969 |
Gene Name: | CD69 molecule |
Gene Aliases: |
AIM, BL-AC/P26, CLEC2C, EA1, GP32/28, MLR-3 |
Location: |
Chr.12:9752486-9760901 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD69 | NM_001781.2 | NP_001772.1 | ||
| AK291869.1 | ||||
| AK303174.1 | ||||
| AK303383.1 | ||||
| AY238518.1 | AAO63584.1 | |||
| BC007037.1 | AAH07037.1 | |||
| L07555.1 | AAB46359.1 | |||
| Z22576.1 | CAA80298.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1051530 | Chr.12:8442286 - 9951690 on Build GRCh38 | Gain |
|
| esv2759878 | Chr.12:9218999 - 9770933 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map