Assay Details
Target Gene Details
Entrez Gene ID: | 171425 |
Gene Name: | citrate lyase beta like |
Gene Aliases: |
CLB |
Location: |
Chr.13:99606664-99909459 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLYBL | NM_206808.3 | NP_996531.1 | ||
| NR_104592.1 | ||||
| XM_005254030.2 | XP_005254087.1 | |||
| XM_005254031.2 | XP_005254088.1 | |||
| XM_006719915.3 | XP_006719978.1 | |||
| XM_011521051.2 | XP_011519353.1 | |||
| XM_011521052.2 | XP_011519354.1 | |||
| XM_017020402.1 | XP_016875891.1 | |||
| XM_017020403.1 | XP_016875892.1 | |||
| XM_017020404.1 | XP_016875893.1 | |||
| AF428253.1 | AAL84703.1 | |||
| AK095506.1 | BAC04561.1 | |||
| AK300510.1 | ||||
| BC034360.1 | AAH34360.1 | |||
| DA621336.1 |
Target Gene Details
Entrez Gene ID: | 101927437 |
Gene Name: | uncharacterized LOC101927437 |
Gene Aliases: |
- |
Location: |
Chr.13:99740870-99957165 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927437 | NR_120421.1 | |||
| DA140610.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv821675 | Chr.13:99790630 - 99832042 on Build GRCh38 | Loss |
|
| esv2747875 | Chr.13:99669768 - 100007232 on Build GRCh38 | Deletion |
|
| nsv832696 | Chr.13:99667339 - 99829982 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map