Assay Details
Target Gene Details
Entrez Gene ID: | 100505996 |
Gene Name: | long intergenic non-protein coding RNA 403 |
Gene Aliases: |
- |
Location: |
Chr.13:111972310-112108015 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC00403 | NR_120392.1 | |||
| DA195709.1 |
Target Gene Details
Entrez Gene ID: | 105378193 |
Gene Name: | serine/arginine repetitive matrix protein 5-like |
Gene Aliases: |
- |
Location: |
Chr.13:112013691-112036593 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105378193 | XM_017020887.1 | XP_016876376.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv351n67 | Chr.13:112033868 - 112116516 on Build GRCh38 | Gain |
|
| nsv563189 | Chr.13:111979751 - 112146300 on Build GRCh38 | Loss |
|
| nsv826807 | Chr.13:112034289 - 112083084 on Build GRCh38 | Gain |
|

Genomic Map