Assay Details
Target Gene Details
Entrez Gene ID: | 100505996 |
Gene Name: | long intergenic non-protein coding RNA 403 |
Gene Aliases: |
- |
Location: |
Chr.13:111972310-112108015 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC00403 | NR_120392.1 | |||
| DA195709.1 |

Genomic Map