Assay Details
Target Gene Details
Entrez Gene ID: | 55270 |
Gene Name: | nudix hydrolase 15 |
Gene Aliases: |
MTH2, NUDT15D |
Location: |
Chr.13:48037567-48052742 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUDT15 | NR_136687.1 | |||
| NR_136688.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832601 | Chr.13:47995817 - 48161803 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78938360] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map