Assay Details
Target Gene Details
Entrez Gene ID: | 22821 |
Gene Name: | RAS p21 protein activator 3 |
Gene Aliases: |
GAP1IP4BP, GAPIII |
Location: |
Chr.13:113977783-114132623 on Build GRCh38 |
Assay Gene Location: | Within Intron 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RASA3 | NM_001320821.1 | NP_001307750.1 | ||
| NM_001320822.1 | NP_001307751.1 | |||
| NM_007368.3 | NP_031394.2 | |||
| XM_011534841.2 | XP_011533143.1 | |||
| XM_011534842.2 | XP_011533144.1 | |||
| XM_017020438.1 | XP_016875927.1 | |||
| XM_017020439.1 | XP_016875928.1 | |||
| AK128094.1 | ||||
| AK289746.1 | ||||
| AK294913.1 | ||||
| AK313795.1 | ||||
| BC038456.1 | AAH38456.1 | |||
| BC047242.1 | AAH47242.1 | |||
| X89399.1 | CAA61580.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv563428 | Chr.13:113970798 - 114076351 on Build GRCh38 | Loss |
|
| esv3633629 | Chr.13:113971705 - 114135826 on Build GRCh38 | Gain |
|
| nsv518916 | Chr.13:113971527 - 114051593 on Build GRCh38 | Loss |
|
| nsv826839 | Chr.13:113970670 - 114166257 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map