Assay Details
Target Gene Details
Entrez Gene ID: | 55208 |
Gene Name: | defective in cullin neddylation 1 domain containing 2 |
Gene Aliases: |
C13orf17 |
Location: |
Chr.13:113455819-113492047 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCUN1D2 | NM_001014283.1 | NP_001014305.1 | ||
| XM_017020643.1 | XP_016876132.1 | |||
| XM_017020644.1 | XP_016876133.1 | |||
| XM_017020645.1 | XP_016876134.1 | |||
| XM_017020646.1 | XP_016876135.1 | |||
| XM_017020647.1 | XP_016876136.1 | |||
| XM_017020648.1 | XP_016876137.1 | |||
| XM_017020649.1 | XP_016876138.1 | |||
| AK000099.1 | BAA90944.1 | |||
| AK001566.1 | BAA91760.1 | |||
| BC056669.1 | AAH56669.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1048814 | Chr.13:113081728 - 113611496 on Build GRCh38 | Gain |
|
| nsv1035248 | Chr.13:113454525 - 113505449 on Build GRCh38 | Loss |
|
| nsv456137 | Chr.13:113452383 - 113477639 on Build GRCh38 | Loss |
|
| esv2759970 | Chr.13:113384114 - 113556749 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map