Assay Details
Target Gene Details
Entrez Gene ID: | 728591 |
Gene Name: | coiled-coil domain containing 169 |
Gene Aliases: |
C13orf38 |
Location: |
Chr.13:36227042-36297855 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC169 | NM_001144981.2 | 8 | 766 | NP_001138453.1 |
| NM_001144982.2 | NP_001138454.1 | |||
| NM_001144983.2 | NP_001138455.1 | |||
| NM_001144984.2 | NP_001138456.1 | |||
| NM_001144985.2 | 7 | 686 | NP_001138457.1 | |
| NM_001144986.2 | 7 | 682 | NP_001138458.1 | |
| NM_001198908.1 | NP_001185837.1 | |||
| AK308999.1 | 7 | 655 | ||
| BC146975.1 | ||||
| BC171867.1 | ||||
| BC171872.1 | ||||
| BC171874.1 | ||||
| BC171880.1 | ||||
| BC171881.1 | ||||
| BC171882.1 | ||||
| BC171884.1 |
Target Gene Details
Entrez Gene ID: | 100526761 |
Gene Name: | CCDC169-SOHLH2 readthrough |
Gene Aliases: |
C13orf38-SOHLH2 |
Location: |
Chr.13:36168208-36297855 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC169-SOHLH2 | NM_001198910.1 | NP_001185839.1 | ||
| AK301863.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3631778 | Chr.13:36226829 - 36237183 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map