Assay Details
Target Gene Details
Entrez Gene ID: | 10186 |
Gene Name: | lipoma HMGIC fusion partner |
Gene Aliases: |
- |
Location: |
Chr.13:39342892-39603219 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LHFP | NM_005780.2 | NP_005771.1 | ||
| XM_011534861.1 | XP_011533163.1 | |||
| AF098807.1 | AAD31386.1 | |||
| AK129819.1 | ||||
| AK223369.1 | BAD97089.1 | |||
| AK313036.1 | ||||
| BC017824.1 | AAH17824.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv820008 | Chr.13:39359864 - 39361437 on Build GRCh38 | Loss |
|
| nsv1036674 | Chr.13:39267345 - 39433980 on Build GRCh38 | Loss |
|
| nsv1054600 | Chr.13:39227146 - 39970890 on Build GRCh38 | Gain |
|
| esv996813 | Chr.13:39355568 - 39362933 on Build GRCh38 | Deletion |
|
| esv25053 | Chr.13:39359490 - 39361250 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map