Assay Details
Target Gene Details
Entrez Gene ID: | 221150 |
Gene Name: | spindle and kinetochore associated complex subunit 3 |
Gene Aliases: |
C13orf3, RAMA1 |
Location: |
Chr.13:21153595-21176602 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SKA3 | NM_001166017.1 | NP_001159489.1 | ||
| NM_145061.5 | NP_659498.4 | |||
| XM_005266288.3 | XP_005266345.1 | |||
| XM_011534994.1 | XP_011533296.1 | |||
| AF361358.1 | AAO13796.1 | |||
| AK090584.1 | BAC03484.1 | |||
| AK307976.1 | ||||
| AK314864.1 | ||||
| BC048988.1 | AAH48988.1 | |||
| BI460315.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1120003 | Chr.13:21168399 - 21172338 on Build GRCh38 | Deletion |
|
| esv3550142 | Chr.13:21168263 - 21172358 on Build GRCh38 | Deletion |
|
| nsv64608 | Chr.13:21168402 - 21172340 on Build GRCh38 | Deletion |
|
| nsv1037082 | Chr.13:20932196 - 21204957 on Build GRCh38 | Gain |
|
| esv3631456 | Chr.13:21168391 - 21176397 on Build GRCh38 | Loss |
|
| nsv1148852 | Chr.13:21168443 - 21172352 on Build GRCh38 | Deletion |
|
| esv3631455 | Chr.13:21168192 - 21172357 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs149604641] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map