Assay Details
Target Gene Details
Entrez Gene ID: | 55270 |
Gene Name: | nudix hydrolase 15 |
Gene Aliases: |
MTH2, NUDT15D |
Location: |
Chr.13:48037567-48052742 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUDT15 | NM_001304745.1 | NP_001291674.1 | ||
| NM_018283.3 | NP_060753.1 | |||
| NR_136687.1 | ||||
| NR_136688.1 | ||||
| AK001818.1 | BAA91925.1 | |||
| BC064607.1 | AAH64607.1 | |||
| BC107875.1 | AAI07876.1 | |||
| BC133015.1 | ||||
| BC133017.1 | ||||
| DA000395.1 | ||||
| DA760263.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832601 | Chr.13:47995817 - 48161803 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map