Assay Details
Target Gene Details
Entrez Gene ID: | 728591 |
Gene Name: | coiled-coil domain containing 169 |
Gene Aliases: |
C13orf38 |
Location: |
Chr.13:36227042-36297855 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC169 | NM_001144981.2 | NP_001138453.1 | ||
| NM_001144982.2 | NP_001138454.1 | |||
| NM_001144983.2 | NP_001138455.1 | |||
| NM_001144984.2 | NP_001138456.1 | |||
| NM_001144985.2 | NP_001138457.1 | |||
| NM_001144986.2 | NP_001138458.1 | |||
| NM_001198908.1 | NP_001185837.1 | |||
| AK308999.1 | ||||
| BC146975.1 | ||||
| BC171867.1 | ||||
| BC171872.1 | ||||
| BC171874.1 | ||||
| BC171880.1 | ||||
| BC171881.1 | ||||
| BC171882.1 | ||||
| BC171884.1 | ||||
| DN999189.1 |
Target Gene Details
Entrez Gene ID: | 100526761 |
Gene Name: | CCDC169-SOHLH2 readthrough |
Gene Aliases: |
C13orf38-SOHLH2 |
Location: |
Chr.13:36168208-36297855 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC169-SOHLH2 | NM_001198910.1 | NP_001185839.1 | ||
| AK301863.1 |
More Information
Set Membership: |
|

Genomic Map