Assay Details
Target Gene Details
Entrez Gene ID: | 55002 |
Gene Name: | transmembrane and coiled-coil domains 3 |
Gene Aliases: |
C13orf11 |
Location: |
Chr.13:113490993-113550229 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMCO3 | NM_017905.4 | NP_060375.4 | ||
| XM_006719969.1 | XP_006720032.1 | |||
| XM_011537498.2 | XP_011535800.1 | |||
| XM_011537501.2 | XP_011535803.1 | |||
| XM_011537502.2 | XP_011535804.1 | |||
| XM_011537504.2 | XP_011535806.1 | |||
| XM_011537506.2 | XP_011535808.1 | |||
| XM_017020629.1 | XP_016876118.1 | |||
| XM_017020630.1 | XP_016876119.1 | |||
| XM_017020631.1 | XP_016876120.1 | |||
| XM_017020632.1 | XP_016876121.1 | |||
| XM_017020633.1 | XP_016876122.1 | |||
| XM_017020634.1 | XP_016876123.1 | |||
| XM_017020635.1 | XP_016876124.1 | |||
| AK000630.1 | BAA91295.1 | |||
| AK074415.1 | BAB85074.1 | |||
| AY358768.1 | AAQ89128.1 | |||
| BC012564.1 | AAH12564.1 | |||
| BC068515.1 | AAH68515.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1048814 | Chr.13:113081728 - 113611496 on Build GRCh38 | Gain |
|
| nsv1053362 | Chr.13:113505488 - 113520943 on Build GRCh38 | Gain |
|
| dgv376e214 | Chr.13:113502377 - 113522478 on Build GRCh38 | Loss |
|
| esv2759970 | Chr.13:113384114 - 113556749 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map