Assay Details
Target Gene Details
Entrez Gene ID: | 23263 |
Gene Name: | MCF.2 cell line derived transforming sequence like |
Gene Aliases: |
ARHGEF14, DBS, OST |
Location: |
Chr.13:112894378-113099739 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MCF2L | XM_011537482.1 | XP_011535784.1 | ||
| XM_011537483.2 | XP_011535785.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759969 | Chr.13:112722543 - 113036887 on Build GRCh38 | Loss |
|
| nsv509518 | Chr.13:112871364 - 112919528 on Build GRCh38 | Insertion |
|
| nsv1047144 | Chr.13:112866048 - 113073300 on Build GRCh38 | Gain |
|
| nsv832723 | Chr.13:112848506 - 113044458 on Build GRCh38 | Loss |
|
| nsv1044644 | Chr.13:112437023 - 113055517 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114701203,rs79487125] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map