Assay Details
Target Gene Details
Entrez Gene ID: | 23250 |
Gene Name: | ATPase phospholipid transporting 11A |
Gene Aliases: |
ATPIH, ATPIS |
Location: |
Chr.13:112690034-112887168 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP11A | NM_015205.2 | NP_056020.2 | ||
| NM_032189.3 | NP_115565.3 | |||
| XM_005268299.4 | XP_005268356.1 | |||
| XM_005268300.4 | XP_005268357.1 | |||
| XM_005268301.4 | XP_005268358.1 | |||
| XM_005268303.4 | XP_005268360.1 | |||
| XM_005268305.4 | XP_005268362.1 | |||
| XM_005268306.4 | XP_005268363.1 | |||
| XM_011537480.2 | XP_011535782.1 | |||
| XM_017020490.1 | XP_016875979.1 | |||
| XM_017020491.1 | XP_016875980.1 | |||
| XM_017020492.1 | XP_016875981.1 | |||
| BE675783.1 |
Target Gene Details
Entrez Gene ID: | 100874205 |
Gene Name: | ATP11A antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.13:112745449-112754731 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP11A-AS1 | NR_046661.1 | |||
| NR_109811.1 | ||||
| AK093292.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759969 | Chr.13:112722543 - 113036887 on Build GRCh38 | Loss |
|
| nsv563230 | Chr.13:112744470 - 112784143 on Build GRCh38 | Gain |
|
| nsv563229 | Chr.13:112685447 - 112791908 on Build GRCh38 | Loss |
|
| nsv518792 | Chr.13:112679370 - 112785272 on Build GRCh38 | Loss |
|
| nsv1044644 | Chr.13:112437023 - 113055517 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map