Assay Details
Target Gene Details
Entrez Gene ID: | 57523 |
Gene Name: | NYN domain and retroviral integrase containing |
Gene Aliases: |
CGIN1, KIAA1305 |
Location: |
Chr.14:24398786-24419288 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NYNRIN | NM_025081.2 | 4 | 1863 | NP_079357.2 |
| AB037726.2 | 2 | 1308 | BAA92543.2 | |
| AK021873.1 | BAB13925.1 | |||
| BX248773.1 | 2 | 870 | CAD66580.1 |
More Information
Set Membership: |
|

Genomic Map