Assay Details
Target Gene Details
Entrez Gene ID: | 51466 |
Gene Name: | Enah/Vasp-like |
Gene Aliases: |
RNB6 |
Location: |
Chr.14:99971454-100144236 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EVL | NM_016337.2 | NP_057421.1 | ||
| XM_005267749.3 | XP_005267806.1 | |||
| XM_011536828.2 | XP_011535130.1 | |||
| XM_017021363.1 | XP_016876852.1 | |||
| XM_017021364.1 | XP_016876853.1 | |||
| AF052504.1 | AAF21709.1 | |||
| AF087843.1 | AAP97156.1 | |||
| AF112209.1 | AAF17197.1 | |||
| AF131766.1 | AAD20040.1 | |||
| AK289720.1 | ||||
| AK295919.1 | ||||
| AK301379.1 | ||||
| BC023997.2 | AAH23997.1 | |||
| BC032358.1 | AAH32358.1 | |||
| BQ083568.1 |
Target Gene Details
Entrez Gene ID: | 442909 |
Gene Name: | microRNA 342 |
Gene Aliases: |
MIRN342, hsa-mir-342 |
Location: |
Chr.14:100109655-100109753 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR342 | NR_029888.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832874 | Chr.14:100068470 - 100218507 on Build GRCh38 | Loss |
|
| nsv527130 | Chr.14:100106617 - 100179544 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map