Assay Details
Target Gene Details
Entrez Gene ID: | 23186 |
Gene Name: | REST corepressor 1 |
Gene Aliases: |
COREST, RCOR |
Location: |
Chr.14:102592638-102730576 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RCOR1 | NM_015156.3 | NP_055971.2 | ||
| AI337324.1 | ||||
| BC051003.1 | AAH51003.1 | |||
| BC064495.1 | AAH64495.1 | |||
| DB455887.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1118294 | Chr.14:102591363 - 102592763 on Build GRCh38 | Deletion |
|
| nsv1049544 | Chr.14:102464997 - 102667048 on Build GRCh38 | Gain |
|
| dgv263n27 | Chr.14:102467010 - 102604508 on Build GRCh38 | Gain |
|
| nsv832880 | Chr.14:102431423 - 102610536 on Build GRCh38 | Loss |
|
| nsv1136183 | Chr.14:102592063 - 102592963 on Build GRCh38 | Deletion |
|
| nsv1046125 | Chr.14:102526629 - 102742816 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map