Assay Details
Target Gene Details
Entrez Gene ID: | 57820 |
Gene Name: | cyclin B1 interacting protein 1 |
Gene Aliases: |
C14orf18, HEI10 |
Location: |
Chr.14:20311368-20333312 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCNB1IP1 | NM_021178.4 | NP_067001.3 | ||
| NM_182849.2 | NP_878269.1 | |||
| NM_182852.3 | NP_878272.1 | |||
| AK026233.1 | BAB15403.1 | |||
| AK225253.1 | ||||
| AK225451.1 | ||||
| AK226037.1 | ||||
| AK309183.1 | ||||
| AL161994.1 | CAB82326.1 | |||
| BI771433.1 | ||||
| BI917081.1 | ||||
| BM741413.1 | ||||
| BX247978.1 | CAD62312.1 | |||
| CB113193.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1042025 | Chr.14:20319802 - 20349692 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map