Assay Details
Target Gene Details
Entrez Gene ID: | 113146 |
Gene Name: | AHNAK nucleoprotein 2 |
Gene Aliases: |
C14orf78 |
Location: |
Chr.14:104937253-104978357 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AHNAK2 | NM_138420.2 | 7 | 17866 | NP_612429.2 |
| XM_005267299.1 | 7 | 17719 | XP_005267356.1 | |
| AB095939.1 | 1 | 8767 | BAC23115.1 | |
| AK095704.1 | 1 | 1666 | ||
| AK126472.1 | 1 | 4348 | ||
| AL834367.1 | 1 | 2910 | CAD39030.1 | |
| BC011859.2 | 1 | 1568 | AAH11859.2 | |
| BC033372.1 | 1 | 1474 | AAH33372.1 | |
| BC049216.1 | 1 | 1908 | AAH49216.1 | |
| BC090889.1 | 3 | 2984 | AAH90889.1 | |
| BX537923.1 | 1 | 3748 | CAD97904.1 | |
| BX538040.1 | 1 | 3522 | CAD97981.1 | |
| BX538106.1 | 1 | 3988 | CAD98019.1 | |
| BX538133.1 | 1 | 3409 | CAD98034.1 |
Target Gene Details
Entrez Gene ID: | 122618 |
Gene Name: | phospholipase D family member 4 |
Gene Aliases: |
C14orf175 |
Location: |
Chr.14:104924806-104937789 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLD4 | XM_011536411.2 | 11 | 2084 | XP_011534713.1 |
| XM_017020965.1 | 11 | 1973 | XP_016876454.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv566099 | Chr.14:104818822 - 104946204 on Build GRCh38 | Gain |
|
| nsv566139 | Chr.14:104880959 - 104950810 on Build GRCh38 | Loss |
|
| esv25274 | Chr.14:104934334 - 104953760 on Build GRCh38 | Gain |
|
| nsv1049253 | Chr.14:104918622 - 105005518 on Build GRCh38 | Gain |
|
| nsv952276 | Chr.14:104922864 - 105021963 on Build GRCh38 | Deletion |
|
| nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79161962] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map