Assay Details
Target Gene Details
Entrez Gene ID: | 10858 |
Gene Name: | cytochrome P450 family 46 subfamily A member 1 |
Gene Aliases: |
CP46, CYP46 |
Location: |
Chr.14:99684262-99727321 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP46A1 | NM_006668.1 | NP_006659.1 | ||
| XM_011536364.1 | XP_011534666.1 | |||
| XM_017020933.1 | XP_016876422.1 | |||
| AB209749.1 | BAD92986.1 | |||
| AF094480.1 | AAD41244.1 | |||
| AK295216.1 | ||||
| BC022539.1 | AAH22539.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv565733 | Chr.14:99522709 - 100097807 on Build GRCh38 | Gain |
|
| esv3635488 | Chr.14:99660725 - 99718739 on Build GRCh38 | Gain |
|
| nsv565763 | Chr.14:99683165 - 99767016 on Build GRCh38 | Loss |
|
| nsv470667 | Chr.14:99658311 - 99742653 on Build GRCh38 | Loss |
|
| nsv1046296 | Chr.14:99640025 - 99723163 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79947893] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map