Assay Details
Target Gene Details
Entrez Gene ID: | 8788 |
Gene Name: | delta like non-canonical Notch ligand 1 |
Gene Aliases: |
DLK, DLK-1, Delta1, FA1, PREF1, Pref-1, ZOG, pG2 |
Location: |
Chr.14:100726865-100738224 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DLK1 | NM_001317172.1 | 6 | 2613 | NP_001304101.1 |
| NM_003836.6 | 5 | 2832 | NP_003827.3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv483116 | Chr.14:100674835 - 100852920 on Build GRCh38 | Gain |
|
| esv992587 | Chr.14:100736422 - 100737196 on Build GRCh38 | Loss |
|
| nsv832876 | Chr.14:100674833 - 100852919 on Build GRCh38 | Loss |
|
| nsv525723 | Chr.14:100634249 - 100737898 on Build GRCh38 | Loss |
|
| esv2751158 | Chr.14:100601952 - 100852910 on Build GRCh38 | Gain |
|
| nsv1425 | Chr.14:100728723 - 100762451 on Build GRCh38 | Insertion |
|
| esv3635516 | Chr.14:100694179 - 100828695 on Build GRCh38 | Gain |
|
| nsv565858 | Chr.14:100722868 - 100766909 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map