Assay Details
Target Gene Details
Entrez Gene ID: | 64150 |
Gene Name: | DIO3 opposite strand/antisense RNA (head to head) |
Gene Aliases: |
C14orf134, DIO3-AS1, DIO3-OS, NCRNA00041 |
Location: |
Chr.14:101552223-101555676 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DIO3OS | NR_002770.1 | 1 | 1298 | |
| AF469201.1 | ||||
| AF469202.1 | 1 | 1298 | ||
| AF469203.1 | ||||
| AF469206.1 | ||||
| AF469207.1 | ||||
| AF469208.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1047869 | Chr.14:101506888 - 101624595 on Build GRCh38 | Gain |
|
| nsv952247 | Chr.14:101543464 - 101564963 on Build GRCh38 | Deletion |
|
| nsv565880 | Chr.14:101539186 - 101625954 on Build GRCh38 | Loss |
|
| dgv1977n100 | Chr.14:101501318 - 101555058 on Build GRCh38 | Gain |
|
| esv3635528 | Chr.14:101459474 - 101559743 on Build GRCh38 | Gain |
|
| dgv1978n100 | Chr.14:101504179 - 101603185 on Build GRCh38 | Gain |
|

Genomic Map