Assay Details
Target Gene Details
Entrez Gene ID: | 22938 |
Gene Name: | SNW domain containing 1 |
Gene Aliases: |
Bx42, NCOA-62, PRPF45, Prp45, SKIIP, SKIP |
Location: |
Chr.14:77717599-77761220 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNW1 | NM_001318844.1 | NP_001305773.1 | ||
| NM_012245.2 | NP_036377.1 | |||
| XM_005267413.3 | XP_005267470.1 | |||
| AF045184.1 | AAC31697.1 | |||
| AK292274.1 | ||||
| AK293619.1 | ||||
| AK302246.1 | ||||
| AK316087.1 | ||||
| AX775981.1 | CAE11690.1 | |||
| BC040112.2 | AAH40112.1 | |||
| BC046105.2 | AAH46105.2 | |||
| BC108903.1 | AAI08904.1 | |||
| BT020060.1 | AAV38863.1 | |||
| BT020061.1 | AAV38864.1 | |||
| CR457433.1 | CAG33714.1 | |||
| U51432.1 | AAC15912.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3635010 | Chr.14:77630281 - 77745656 on Build GRCh38 | Gain |
|
| nsv520021 | Chr.14:77686578 - 77741951 on Build GRCh38 | Gain |
|
| esv34940 | Chr.14:77638554 - 77756455 on Build GRCh38 | Gain |
|
| nsv521287 | Chr.14:77641660 - 77744882 on Build GRCh38 | Gain |
|
| esv2759998 | Chr.14:77607837 - 77796229 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map