Assay Details
Target Gene Details
Entrez Gene ID: | 84439 |
Gene Name: | HHIP like 1 |
Gene Aliases: |
KIAA1822, UNQ9245 |
Location: |
Chr.14:99604476-99680569 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HHIPL1 | XM_006720277.3 | XP_006720340.1 | ||
| XM_011537236.2 | XP_011535538.1 | |||
| XM_017021708.1 | XP_016877197.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1071167 | Chr.14:99603062 - 99604863 on Build GRCh38 | Deletion |
|
| nsv952236 | Chr.14:99504664 - 99604963 on Build GRCh38 | Deletion |
|
| dgv3892n54 | Chr.14:99603237 - 99616411 on Build GRCh38 | Loss |
|
| nsv565747 | Chr.14:99603083 - 99606662 on Build GRCh38 | Gain+Loss |
|
| nsv565735 | Chr.14:99589385 - 99683165 on Build GRCh38 | Loss |
|
| nsv565733 | Chr.14:99522709 - 100097807 on Build GRCh38 | Gain |
|
| dgv3887n54 | Chr.14:99602384 - 99606662 on Build GRCh38 | Loss |
|
| nsv827051 | Chr.14:99602761 - 99605506 on Build GRCh38 | Gain |
|
| nsv827050 | Chr.14:99599494 - 99608837 on Build GRCh38 | Gain |
|
| dgv3891n54 | Chr.14:99603186 - 99609119 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map