Assay Details
Target Gene Details
Entrez Gene ID: | 57523 |
Gene Name: | NYN domain and retroviral integrase containing |
Gene Aliases: |
CGIN1, KIAA1305 |
Location: |
Chr.14:24398786-24419288 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NYNRIN | NM_025081.2 | NP_079357.2 | ||
| AK021873.1 | BAB13925.1 | |||
| BC065283.1 | AAH65283.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs75391305] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map