Assay Details
Target Gene Details
Entrez Gene ID: | 51466 |
Gene Name: | Enah/Vasp-like |
Gene Aliases: |
RNB6 |
Location: |
Chr.14:99971454-100144236 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EVL | XM_017021364.1 | XP_016876853.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv565733 | Chr.14:99522709 - 100097807 on Build GRCh38 | Gain |
|
| nsv1112652 | Chr.14:99971363 - 99972063 on Build GRCh38 | Deletion |
|
| nsv1071169 | Chr.14:99971362 - 99972463 on Build GRCh38 | Deletion |
|
| nsv832873 | Chr.14:99820652 - 99997237 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map