Assay Details
Target Gene Details
Entrez Gene ID: | 204219 |
Gene Name: | ceramide synthase 3 |
Gene Aliases: |
ARCI9, LASS3 |
Location: |
Chr.15:100400395-100544982 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CERS3 | NM_001290341.2 | NP_001277270.1 | ||
| NM_001290342.2 | NP_001277271.1 | |||
| NM_001290343.1 | NP_001277272.1 | |||
| NM_178842.4 | NP_849164.2 | |||
| XM_011521355.2 | XP_011519657.1 | |||
| XM_011521357.2 | XP_011519659.1 | |||
| XM_011521358.1 | XP_011519660.1 | |||
| XM_017022002.1 | XP_016877491.1 | |||
| XM_017022003.1 | XP_016877492.1 | |||
| XM_017022004.1 | XP_016877493.1 | |||
| BC028703.1 | AAH28703.1 | |||
| BC034500.1 | AAH34500.1 | |||
| BC034970.1 | AAH34970.1 | |||
| HY237628.1 |
Target Gene Details
Entrez Gene ID: | 102723320 |
Gene Name: | CERS3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.15:100372939-100437914 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CERS3-AS1 | NR_120374.1 | |||
| AI160488.1 | ||||
| AL708976.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3569898 | Chr.15:100394057 - 100550681 on Build GRCh38 | Gain |
|
| esv3637473 | Chr.15:100311230 - 100508356 on Build GRCh38 | Gain |
|
| nsv1036363 | Chr.15:100389827 - 100434244 on Build GRCh38 | Loss |
|
| esv3637474 | Chr.15:100317437 - 100437921 on Build GRCh38 | Gain |
|
| nsv471265 | Chr.15:100296251 - 100584877 on Build GRCh38 | Gain |
|
| esv3892745 | Chr.15:100226796 - 100468465 on Build GRCh38 | Gain |
|
| nsv457284 | Chr.15:100201823 - 100595854 on Build GRCh38 | Gain |
|
| nsv1045838 | Chr.15:99702862 - 100492013 on Build GRCh38 | Gain |
|
| nsv457287 | Chr.15:100299689 - 100413206 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map