Assay Details
Target Gene Details
Entrez Gene ID: | 83440 |
Gene Name: | ADP dependent glucokinase |
Gene Aliases: |
2610017G09Rik, ADP-GK |
Location: |
Chr.15:72751367-72783785 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADPGK | NM_031284.4 | NP_112574.3 | ||
| NR_023318.1 | ||||
| NR_023319.1 | ||||
| XM_006720712.3 | XP_006720775.1 | |||
| XM_006720713.3 | XP_006720776.1 | |||
| XM_011522102.2 | XP_011520404.1 | |||
| XM_011522103.2 | XP_011520405.1 | |||
| XM_011522104.2 | XP_011520406.1 | |||
| AF204270.1 | AAL31473.1 | |||
| AK055526.1 | BAB70941.1 | |||
| AK075560.1 | BAC11699.1 | |||
| AK095995.1 | ||||
| AK293282.1 | ||||
| AK294391.1 | ||||
| BC006112.1 | AAH06112.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2760039 | Chr.15:72527444 - 72828219 on Build GRCh38 | Gain+Loss |
|
| nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map