Assay Details
Target Gene Details
Entrez Gene ID: | 102723320 |
Gene Name: | CERS3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.15:100372939-100437914 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CERS3-AS1 | NR_120374.1 | |||
| AL708976.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3637473 | Chr.15:100311230 - 100508356 on Build GRCh38 | Gain |
|
| nsv1036363 | Chr.15:100389827 - 100434244 on Build GRCh38 | Loss |
|
| esv3637474 | Chr.15:100317437 - 100437921 on Build GRCh38 | Gain |
|
| nsv471265 | Chr.15:100296251 - 100584877 on Build GRCh38 | Gain |
|
| esv3892745 | Chr.15:100226796 - 100468465 on Build GRCh38 | Gain |
|
| nsv457284 | Chr.15:100201823 - 100595854 on Build GRCh38 | Gain |
|
| nsv1045838 | Chr.15:99702862 - 100492013 on Build GRCh38 | Gain |
|
| nsv457287 | Chr.15:100299689 - 100413206 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77445763] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map