Assay Details
Target Gene Details
Entrez Gene ID: | 100505648 |
Gene Name: | RAD51 antisense RNA 1 (head to head) |
Gene Aliases: |
TODRA |
Location: |
Chr.15:40693734-40695105 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAD51-AS1 | NR_040058.1 | 2 | 288 | |
| AK125393.1 | 2 | 288 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2760026 | Chr.15:40540346 - 40702913 on Build GRCh38 | Loss |
|
| nsv832984 | Chr.15:40502192 - 40706337 on Build GRCh38 | Loss |
|
| nsv529030 | Chr.15:40664273 - 40720497 on Build GRCh38 | Loss |
|

Genomic Map