Assay Details
Target Gene Details
Entrez Gene ID: | 6638 |
Gene Name: | small nuclear ribonucleoprotein polypeptide N |
Gene Aliases: |
HCERN3, PWCR, RT-LI, SM-D, SMN, SNRNP-N, SNURF-SNRPN, sm-N |
Location: |
Chr.15:24823637-24978723 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNRPN | NM_022806.3 | NP_073717.1 | ||
| NM_022807.3 | NP_073718.1 | |||
| NM_022808.3 | NP_073719.1 | |||
| DA069028.1 | ||||
| DA131564.1 | ||||
| L80005.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1045067 | Chr.15:24792359 - 25083568 on Build GRCh38 | Gain |
|
| nsv568530 | Chr.15:24540053 - 24862075 on Build GRCh38 | Loss |
|
| nsv568533 | Chr.15:24843068 - 24861936 on Build GRCh38 | Loss |
|
| nsv9198 | Chr.15:24852579 - 24855381 on Build GRCh38 | Loss |
|
| esv3581563 | Chr.15:24843286 - 24874807 on Build GRCh38 | Loss |
|
| esv2760019 | Chr.15:24810131 - 24892816 on Build GRCh38 | Loss |
|
| nsv516596 | Chr.15:24847119 - 24869979 on Build GRCh38 | Loss |
|
| nsv456730 | Chr.15:24821213 - 24869979 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74005377] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map