Assay Details
Target Gene Details
Entrez Gene ID: | 342125 |
Gene Name: | transmembrane channel like 3 |
Gene Aliases: |
- |
Location: |
Chr.15:81332419-81374077 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMC3 | NM_001080532.1 | NP_001074001.1 | ||
| AY236490.1 | AAP69868.1 | |||
| AY263163.1 | AAP78778.1 | |||
| BC132679.1 | ||||
| BC143410.1 | ||||
| BC157874.1 |
Target Gene Details
Entrez Gene ID: | 101929655 |
Gene Name: | TMC3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.15:81324333-81443076 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMC3-AS1 | NR_120365.1 | |||
| BI821075.1 | ||||
| CB134362.1 | ||||
| HY040505.1 | ||||
| HY112626.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1048788 | Chr.15:81257750 - 81343926 on Build GRCh38 | Loss |
|
| nsv1048106 | Chr.15:81325867 - 81344862 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map