Assay Details
Target Gene Details
Entrez Gene ID: | 1198 |
Gene Name: | CDC like kinase 3 |
Gene Aliases: |
PHCLK3, PHCLK3/152 |
Location: |
Chr.15:74598476-74645418 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLK3 | NM_001130028.1 | 1 | 301 | NP_001123500.1 |
| NM_003992.4 | NP_003983.2 | |||
| XM_005254151.3 | 1 | 301 | XP_005254208.1 | |
| XM_011521205.2 | 1 | 301 | XP_011519507.1 | |
| XM_011521206.2 | 1 | 301 | XP_011519508.1 | |
| XM_011521209.2 | 1 | 301 | XP_011519511.2 | |
| XM_017021905.1 | 1 | 301 | XP_016877394.1 | |
| XM_017021906.1 | 1 | 301 | XP_016877395.1 | |
| XM_017021907.1 | 1 | 301 | XP_016877396.1 | |
| XM_017021908.1 | XP_016877397.1 | |||
| XM_017021909.1 | XP_016877398.1 | |||
| XM_017021910.1 | XP_016877399.1 | |||
| AK091692.1 | 1 | 301 | ||
| L29217.1 | AAA61484.1 | |||
| L29220.1 | AAA61483.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv457204 | Chr.15:74592750 - 74655722 on Build GRCh38 | Loss |
|
| nsv817698 | Chr.15:74596822 - 74727108 on Build GRCh38 | Gain |
|
| nsv951994 | Chr.15:74614960 - 74633959 on Build GRCh38 | Deletion |
|
| nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map