Assay Details
Target Gene Details
Entrez Gene ID: | 9051 |
Gene Name: | proline-serine-threonine phosphatase interacting protein 1 |
Gene Aliases: |
CD2BP1, CD2BP1L, CD2BP1S, H-PIP, PAPAS, PSTPIP |
Location: |
Chr.15:76994680-77037475 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSTPIP1 | NM_001321135.1 | NP_001308064.1 | ||
| NM_001321136.1 | NP_001308065.1 | |||
| NM_001321137.1 | NP_001308066.1 | |||
| NM_003978.4 | NP_003969.2 | |||
| NR_135552.1 | ||||
| XM_006720737.2 | XP_006720800.1 | |||
| XM_011522163.1 | XP_011520465.1 | |||
| XM_011522164.1 | XP_011520466.1 | |||
| XM_011522166.1 | XP_011520468.1 | |||
| XM_011522167.1 | XP_011520469.1 | |||
| XM_011522168.2 | XP_011520470.1 | |||
| XM_011522169.1 | XP_011520471.1 | |||
| AB451310.1 | ||||
| AB451440.1 | ||||
| AF038602.1 | AAD11958.1 | |||
| AF038603.1 | AAD11959.1 | |||
| AK298733.1 | ||||
| AK304052.1 | ||||
| AK308462.1 | ||||
| AK309856.1 | ||||
| AK310072.1 | ||||
| AK310126.1 | ||||
| AK316220.1 | ||||
| BC008602.1 | AAH08602.1 | |||
| DA952061.1 | ||||
| DB123759.1 | ||||
| KU178458.1 | ||||
| KU178459.1 | ||||
| U94778.1 | AAD00762.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv517068 | Chr.15:76962203 - 77088880 on Build GRCh38 | Gain+Loss |
|
| esv2760041 | Chr.15:76878209 - 77081106 on Build GRCh38 | Gain |
|
| nsv1043017 | Chr.15:76974435 - 77017597 on Build GRCh38 | Gain |
|
| nsv833058 | Chr.15:76943554 - 77101298 on Build GRCh38 | Loss |
|
| esv2749898 | Chr.15:75764003 - 77933523 on Build GRCh38 | Deletion |
|
| nsv1048833 | Chr.15:77002252 - 77046918 on Build GRCh38 | Gain |
|
| nsv1040852 | Chr.15:76867896 - 77845199 on Build GRCh38 | Gain |
|
| nsv509576 | Chr.15:76962019 - 77047336 on Build GRCh38 | Insertion |
|
| nsv570111 | Chr.15:76994352 - 77040265 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map