Assay Details
Target Gene Details
Entrez Gene ID: | 10391 |
Gene Name: | coronin 2B |
Gene Aliases: |
CLIPINC |
Location: |
Chr.15:68518373-68727806 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CORO2B | NM_001324014.1 | NP_001310943.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1050575 | Chr.15:68482000 - 68554748 on Build GRCh38 | Gain |
|
| nsv529008 | Chr.15:68493946 - 68536147 on Build GRCh38 | Loss |
|
| nsv516489 | Chr.15:68513090 - 68536147 on Build GRCh38 | Gain+Loss |
|
| esv2760036 | Chr.15:68245734 - 68522324 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map