Assay Details
Target Gene Details
Entrez Gene ID: | 23005 |
Gene Name: | mitogen-activated protein kinase binding protein 1 |
Gene Aliases: |
JNKBP-1, JNKBP1 |
Location: |
Chr.15:41774434-41827855 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAPKBP1 | NM_001128608.1 | NP_001122080.1 | ||
| NM_001265611.1 | NP_001252540.1 | |||
| NM_014994.2 | NP_055809.2 | |||
| NR_049761.1 | ||||
| NR_049762.1 | ||||
| XM_006720438.1 | XP_006720501.1 | |||
| XM_011521383.1 | XP_011519685.1 | |||
| XM_011521384.2 | XP_011519686.1 | |||
| XM_017022017.1 | XP_016877506.1 | |||
| AB011168.2 | ||||
| AK091616.1 | ||||
| AK292414.1 | ||||
| AK302482.1 | ||||
| AL833267.1 | ||||
| BC113983.1 | ||||
| BC114493.1 | ||||
| BX640787.1 | ||||
| BX648786.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1038894 | Chr.15:41745954 - 41785254 on Build GRCh38 | Gain |
|
| dgv449e214 | Chr.15:41758480 - 41824304 on Build GRCh38 | Gain |
|
| dgv2581n100 | Chr.15:41712764 - 41806698 on Build GRCh38 | Gain |
|
| esv2760027 | Chr.15:41754975 - 41830419 on Build GRCh38 | Loss |
|
| nsv1049514 | Chr.15:41666284 - 41789012 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map