Assay Details
Target Gene Details
Entrez Gene ID: | 5371 |
Gene Name: | promyelocytic leukemia |
Gene Aliases: |
MYL, PP8675, RNF71, TRIM19 |
Location: |
Chr.15:73994673-74047819 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv457203 | Chr.15:74016599 - 74055382 on Build GRCh38 | Loss |
|
| nsv469619 | Chr.15:73999518 - 74173980 on Build GRCh38 | Loss |
|
| nsv833052 | Chr.15:73958037 - 74084809 on Build GRCh38 | Gain+Loss |
|
| nsv509575 | Chr.15:74018397 - 74102404 on Build GRCh38 | Insertion |
|
| nsv518316 | Chr.15:73998652 - 74038742 on Build GRCh38 | Gain |
|
| esv2760040 | Chr.15:73922773 - 74172575 on Build GRCh38 | Gain+Loss |
|
| nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs117524612] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map