Assay Details
Target Gene Details
Entrez Gene ID: | 729911 |
Gene Name: | ANKRD34C antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.15:79191707-79283945 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD34C-AS1 | NR_038997.1 | |||
| DA316055.1 | ||||
| DA323807.1 | ||||
| DA336675.1 | ||||
| DA364073.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2760042 | Chr.15:79062895 - 79226427 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs138962589,rs78990609] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map