Assay Details
Target Gene Details
Entrez Gene ID: | 342125 |
Gene Name: | transmembrane channel like 3 |
Gene Aliases: |
- |
Location: |
Chr.15:81332419-81374077 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMC3 | NM_001080532.1 | NP_001074001.1 | ||
| AY236490.1 | AAP69868.1 | |||
| AY263163.1 | AAP78778.1 | |||
| BC132679.1 | ||||
| BC143410.1 | ||||
| BC157874.1 |
Target Gene Details
Entrez Gene ID: | 101929655 |
Gene Name: | TMC3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.15:81324333-81443076 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMC3-AS1 | NR_120365.1 | |||
| BI821075.1 | ||||
| CB134362.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs150054745] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map