Assay Details
Target Gene Details
Entrez Gene ID: | 10169 |
Gene Name: | small EDRK-rich factor 2 |
Gene Aliases: |
4F5REL, FAM2C, H4F5REL, HsT17089 |
Location: |
Chr.15:43777096-43796089 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERF2 | NM_001199877.1 | NP_001186806.1 | ||
| BQ061048.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1036318 | Chr.15:43761419 - 43868575 on Build GRCh38 | Gain |
|
| esv2760028 | Chr.15:43530960 - 43856443 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map