Assay Details
Target Gene Details
Entrez Gene ID: | 390616 |
Gene Name: | ankyrin repeat domain 34C |
Gene Aliases: |
- |
Location: |
Chr.15:79282804-79298239 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD34C | NM_001146341.1 | 1 | 227 | NP_001139813.1 |
| AK127037.1 | 1 | 227 |
Target Gene Details
Entrez Gene ID: | 729911 |
Gene Name: | ANKRD34C antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.15:79191707-79283945 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD34C-AS1 | NR_038997.1 | |||
| DA316055.1 | ||||
| DA323807.1 | ||||
| DA336675.1 | ||||
| DA364073.1 |
More Information
Set Membership: |
|

Genomic Map