Assay Details
Target Gene Details
Entrez Gene ID: | 145694 |
Gene Name: | uncharacterized LOC145694 |
Gene Aliases: |
- |
Location: |
Chr.15:69396904-69415029 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC145694 | NR_132970.1 | |||
| NR_132971.1 | ||||
| AK056793.1 | ||||
| CN360125.1 |
Target Gene Details
Entrez Gene ID: | 54852 |
Gene Name: | progestin and adipoQ receptor family member 5 |
Gene Aliases: |
MPRG |
Location: |
Chr.15:69284134-69407637 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAQR5 | NM_001104554.1 | NP_001098024.1 | ||
| NM_017705.3 | NP_060175.3 | |||
| XM_005254495.4 | XP_005254552.1 | |||
| XM_011521720.2 | XP_011520022.1 | |||
| XM_011521722.2 | XP_011520024.1 | |||
| XM_011521723.2 | XP_011520025.1 | |||
| XM_011521725.2 | XP_011520027.1 | |||
| XM_011521727.2 | XP_011520029.1 | |||
| XM_017022360.1 | XP_016877849.1 | |||
| XM_017022361.1 | XP_016877850.1 | |||
| XM_017022362.1 | XP_016877851.1 | |||
| XM_017022363.1 | XP_016877852.1 | |||
| XM_017022364.1 | XP_016877853.1 | |||
| XM_017022366.1 | XP_016877855.1 | |||
| XM_017022368.1 | XP_016877857.1 | |||
| AK000197.1 | BAA91004.1 | |||
| AY424283.1 | AAR08371.1 | |||
| BC039234.1 | AAH39234.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2760037 | Chr.15:69342413 - 69499860 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map