Assay Details
Target Gene Details
Entrez Gene ID: | 5371 |
Gene Name: | promyelocytic leukemia |
Gene Aliases: |
MYL, PP8675, RNF71, TRIM19 |
Location: |
Chr.15:73994673-74047819 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PML | NM_002675.3 | 1 | 51 | NP_002666.1 |
| NM_033238.2 | 1 | 51 | NP_150241.2 | |
| NM_033239.2 | 1 | 51 | NP_150242.1 | |
| NM_033240.2 | 1 | 51 | NP_150243.2 | |
| NM_033244.3 | 1 | 51 | NP_150247.2 | |
| NM_033246.2 | 1 | 51 | NP_150249.1 | |
| NM_033247.2 | 1 | 51 | NP_150250.2 | |
| NM_033249.2 | 1 | 51 | NP_150252.1 | |
| NM_033250.2 | 1 | 51 | NP_150253.2 | |
| AB209411.1 | BAD92648.1 | |||
| AF230401.1 | AAG50180.1 | |||
| AF230402.1 | AAG50181.1 | |||
| AF230403.1 | AAG50182.1 | |||
| AF230404.1 | AAG50183.1 | |||
| AF230405.1 | AAG50184.1 | |||
| AF230406.1 | AAG50185.1 | |||
| AF230407.1 | AAG50186.1 | |||
| AF230408.1 | AAG50187.1 | |||
| AF230409.1 | AAG50188.1 | |||
| AF230410.1 | AAG50189.1 | |||
| AF230411.1 | AAG50190.1 | |||
| BC000080.2 | AAH00080.2 | |||
| BC020994.2 | AAH20994.1 | |||
| M73778.1 | AAA60125.1 | |||
| S50913.1 | AAB19601.2 | |||
| X63131.1 | 1 | 52 | CAA44841.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833052 | Chr.15:73958037 - 74084809 on Build GRCh38 | Gain+Loss |
|
| esv2760040 | Chr.15:73922773 - 74172575 on Build GRCh38 | Gain+Loss |
|
| nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map